missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Saponin permeating solution, 0.5% w/v soln. in PBS (10X), Thermo Scientific Chemicals
Cell permeabilizing agent
Supplier: Thermo Scientific Chemicals J63209AK
Description
Saponin permeating solution, is used as a cell permeabilizing agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Saponin permeating solution | |
Difficult to mix. | |
Liquid |
0.5% w/v soln. in PBS (10X) | |
250 mL |
RUO – Research Use Only