Please login to your online account to display your discounted pricing

SANT-1, Tocris

Inhibitor of hedgehog (Hh) signaling; antagonizes smoothened activity

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material SANT-1
Assay Percent Range >99%
Molecular Formula C23H27N5
Formula Weight 373.5
Synonym N-[(3, 5-dimethyl-1-phenyl-1H-pyrazol-4-yl)methylene]-4-(phenylmethyl)-1-piperazinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

197450

 
r & d systems
197450
50mg Each for $904.70

197410

 
r & d systems
197410
10mg Each for $221.01
Description & Specifications

Specifications

Formula Weight 373.5
Quantity 50mg
Chemical Name or Material SANT-1
CAS 304909-07-7
Synonym N-[(3, 5-dimethyl-1-phenyl-1H-pyrazol-4-yl)methylene]-4-(phenylmethyl)-1-piperazinamine
Assay Percent Range >99%
Molecular Formula C23H27N5
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C