missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sand, Standard, EMD Millipore

$839.57 - $839.57

Chemical Identifiers

CAS 14808-60-7
Physical Form Granular
Molecular Formula SiO2
Linear Formula SiO2
Formula Weight 6.8
Chemical Name or Material Sand, Standard
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MSX007530 Emd Millipore Corporation
SX007530
12kg Each for $839.57
Specifications Description & Specifications

Chemical Identifiers

14808-60-7
SiO2
6.8
Granular
SiO2
Sand, Standard

Specifications

6.8
Beige
14808-60-7
SiO2
12kg
Granular
SiO2
Sand, Standard

Safety and Handling

Recommended Storage : Room Temperature