missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sand, pure, 40-100 mesh, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 370940050

 View more versions of this product

Catalog No. AC370940050



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Filter aid
Specifications

Specifications

2230.0°C
1610.0°C
Beige to Gray
Crystals
O2Si
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
dioxosilane
24261
60.08
Plastic bottle
Sand
40 to 100 Mesh
14808-60-7
5 kg
SiO2
13,8567
Solubility in water: insoluble.
O=[Si]=O
60.08
CHEBI:30563
Pure

Chemical Identifiers

14808-60-7
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
O=[Si]=O
O2Si
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
CHEBI:30563
Safety and Handling

Safety and Handling

EINECSNumber : 238-878-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only