Thermo Scientific™ Sample Storage Assembled Screw Vial Kits
Store your samples with confidence and convenience with these assembled screw vial kits.
$203.85 - $302.64
Specifications
Closure Color White
Closure Material Polypropylene
Material Glass
No. per Pack 100
Description Screw Vial Assembled Kit, Solid-top Cap
View More Specs
Products 2
Catalog Number Mfr. No. Color Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Color Volume (Metric) Price Quantity  
03-375-25
View Documents Promotion Details
Thermo Scientific™
B780020
Clear 20 mL
Pack of 100 Each for $203.85
03-341-120
View Documents Promotion Details
Thermo Scientific™
B780020A
Amber 20 mL
Pack of 100 Each for $302.64
Description
Description
Store your samples with confidence in convenient Thermo Scientific™ National™ Sample Storage Assembled Screw Vial Kits.
  • Superior quality 33 expansion borosilicate clear (Type 1, Class A) or 51A amber (Type 1, Class B) glass vials
  • PTFE-lined solid-top storage caps
  • PTFE on film/foam backing offers broad chemical resistance
  • Not autoclavable
  • Pack of 100
Specifications
Specifications
White
Glass
Screw Vial Assembled Kit, Solid-top Cap
Polypropylene
100
SDS
Product Certifications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.