missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Samarium ingot, 99.9% (REO), Thermo Scientific Chemicals
$219.96 - $613.59
Chemical Identifiers
CAS | 7440-19-9 |
---|---|
Molecular Formula | Sm |
Molecular Weight (g/mol) | 150.36 |
MDL Number | MFCD00011233 MFCD00151299 |
InChI Key | KZUNJOHGWZRPMI-UHFFFAOYSA-N |
Synonym | samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l |
PubChem CID | 23951 |
ChEBI | CHEBI:33374 |
IUPAC Name | samarium |
SMILES | [Sm] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-19-9 | |
150.36 | |
KZUNJOHGWZRPMI-UHFFFAOYSA-N | |
23951 | |
samarium |
Sm | |
MFCD00011233 MFCD00151299 | |
samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l | |
CHEBI:33374 | |
[Sm] |
Specifications
Silver to Gray | |
Silver Gray solid | |
99.9% (REO) | |
MFCD00011233 MFCD00151299 | |
samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l | |
[Sm] | |
150.36 | |
CHEBI:33374 | |
Air-stable poly bottle |
7440-19-9 | |
7.52 g/cm3 | |
Sm | |
25 g | |
KZUNJOHGWZRPMI-UHFFFAOYSA-N | |
samarium | |
23951 | |
Odorless | |
Samarium, ingot |
Safety and Handling
P210-P223-P231+P232-P240-P241-P280-P335+P334-P370+P378q-P501c
H228-H260
DOTInformation : Hazard Class: 4.3; Packaging Group: III
EINECSNumber : 231-128-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only