Please login to your online account to display your discounted pricing

Alfa Aesar™ Samarium powder, <250 micron, REacton™, 99.9% (REO)

$97.74 - $97.74

Chemical Identifiers

Chemical Name or Material Samarium powder
CAS 7440-19-9
Purity Grade Reacton™
MDL Number MFCD00011233
UN Number UN3089
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1242206

 
alfa aesar™
1242206
5g Each for $97.74
Description & Specifications

Specifications

Quantity 5g
Packaging Ampule
Chemical Name or Material Samarium powder
Odor Odorless
TSCA TSCA
Purity Grade Reacton™
Physical Form Powder
Assay 99.9%
Assay Percent Notes (REO)
MDL Number MFCD00011233
CAS 7440-19-9
UN Number UN3089
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
Health Hazard 3 GHS P Statement
P210-P231+P232-P280-P370+P378b-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: Special powder for metal fires.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1
  • Ampouled under argon
DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
Particle Size <250 Micron
Color Gray to silver

  • Ampouled under argon

Safety and Handling

Danger

GHS P Statement
P210-P231+P232-P280-P370+P378b-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: Special powder for metal fires.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

  • Ampouled under argon