Please login to your online account to display your discounted pricing

Alfa Aesar™ Samarium(III) trifluoromethanesulfonate, 98%

$68.42 - $271.22

Chemical Identifiers

Chemical Name or Material Samarium(III) trifluoromethanesulfonate
CAS 52093-28-4
Molecular Formula Sm(SO3CF3)3
Formula Weight 597.56
MDL Number MFCD00192431
Synonym Samarium(III) triflate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2025306

  SDS
alfa aesar™
L2025306
5g Each for $68.42

AAL2025314

  SDS
alfa aesar™
L2025314
25g Each for $271.22
Description & Specifications

Specifications

Formula Weight 597.56
MDL Number MFCD00192431
CAS 52093-28-4
Assay 98%
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula Sm(SO3CF3)3
Synonym Samarium(III) triflate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Chemical Name or Material Samarium(III) trifluoromethanesulfonate

Applications
Used as catalyst for Baylis-Hillman reaction.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store in a cool, dry place in a tightly closed containers. Stable under recommended storage conditions. Incompatible with oxidizing agents and moisture.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.