missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Samarium(III) sulfate octahydrate, REacton™, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1460122

 View more versions of this product

Catalog No. AA1460122



Specifications

Specifications

Samarium(III) sulfate octahydrate
(REO)
14,8347
733.00 (588.88 Anhydrous)
99.9%
450°C (in -8H2O)
13465-58-2
MFCD00011236
Sparingly soluble in water
Crystalline
Reacton™
100g

Chemical Identifiers

13465-58-2
MFCD00011236
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 237-211-4

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents