missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Samarium(III) oxide, REacton™, 99.998% (metals basis), REM 100ppm

$95.47 - $867.89

Chemical Identifiers

CAS 12060-58-1
MDL Number MFCD00011237
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1091506
View Documents
Alfa Aesar™
1091506
5g Each for $95.47
 
AA1091514
View Documents
Alfa Aesar™
1091514
25g Each for $331.08
 
AA1091522
View Documents
Alfa Aesar™
1091522
100g Each for $867.89
 
Specifications

Chemical Identifiers

12060-58-1
MFCD00011237

Specifications

12060-58-1
14,8347
348.7
99.998%
1.97
5g
(metals basis)
MFCD00011237
Insoluble in water.
Powder
Reacton™
2300°C
Samarium(III) oxide, REM 100ppm
Safety and Handling

Safety and Handling

EINECSNumber : 235-043-6

TSCA : Yes

Recommended Storage : Ambient temperatures