missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Samarium(III) oxide, REacton™, 99.998% (metals basis), REM 100ppm

Manufacturer:  Alfa Aesar™ 1091514

 View more versions of this product

Catalog No. AA1091514



Specifications

Specifications

Samarium(III) oxide
12060-58-1
MFCD00011237
348.7
99.998%
1.97
25g
REM 100ppm
(metals basis)
14,8347
Powder
Reacton™
2300°C

Chemical Identifiers

12060-58-1
MFCD00011237
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents