missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Samarium(III) oxide, 99.99% (REO), Alfa Aesar™, Reacton™

Manufacturer:  Alfa Aesar 1123014

 View more versions of this product

Catalog No. AA1123014



Specifications

Specifications

Samarium(III) oxide
99.99% (REO)
MFCD00011237
348.7
Violet
12060-58-1
Sm2O3
Off-white powder
Powder
25g

Chemical Identifiers

12060-58-1
Sm2O3
MFCD00011237
Safety and Handling

Safety and Handling

EINECSNumber : 235-043-6

Documents