missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Samarium(III) oxide, 99.99% (REO), Alfa Aesar™, REacton™

$153.64 - $1,261.65

Chemical Identifiers

CAS 12060-58-1
Molecular Formula Sm2O3
MDL Number MFCD00011237
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1123014
View Documents
Thermo Fisher Scientific
1123014
25g
Each for $153.64
 
AA1123022
View Documents
Thermo Fisher Scientific
1123022
100g
Each for $353.58
 
AA1123036
View Documents
Thermo Fisher Scientific
1123036
500g
Each for $1,261.65
 
Description

Description

Specifications

Chemical Identifiers

12060-58-1
Sm2O3
MFCD00011237

Specifications

12060-58-1
Sm2O3
Off-white powder
348.7
1.97
2300°C
Samarium(III) oxide
99.99% (REO)
MFCD00011237
Insoluble in water.
Powder
Violet
25g
Safety and Handling

Safety and Handling

EINECSNumber : 235-043-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS