missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Samarium(III) oxide, 99.9% (metals basis), Alfa Aesar™

$93.57 - $382.50

Chemical Identifiers

CAS 12060-58-1
Molecular Formula Sm2O3
MDL Number MFCD00011237
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3627809
View Documents
Thermo Fisher Scientific
3627809
10g Each for $93.57
 
AA3627818
View Documents
Thermo Fisher Scientific
3627818
50g Each for $382.50
 
Specifications

Chemical Identifiers

12060-58-1
Sm2O3
MFCD00011237

Specifications

12060-58-1
Sm2O3
Off-white 3-12mm Sintered Lump
348.7
1.97
2300°C
Samarium(III) oxide
99.95% (metals basis)
MFCD00011237
Insoluble in water
3 to 12mm Sintered Lump
White
10g
Safety and Handling

Safety and Handling

EINECSNumber : 235-043-6

TSCA : Yes

Recommended Storage : Ambient temperatures