missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Samarium(III) oxalate hydrate, REacton™, 99.97% (REO)

$126.65 - $403.41

Chemical Identifiers

CAS 14175-03-2
Molecular Formula C6H26O22Sm2
Molecular Weight (g/mol) 750.972
MDL Number MFCD00150489
InChI Key IKPDQYODFRQYAD-UHFFFAOYSA-N
Synonym ethanedioic acid,samarium 3+ salt, hydrate 3:2:10, samarium iii oxalate hydrate 2:3:10
PubChem CID 121488066
IUPAC Name oxalic acid;samarium;decahydrate
SMILES C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.O.O.O.O.O.O.O.O.O.O.[Sm].[Sm]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1317614
View Documents
Alfa Aesar™
1317614
25g
Each for $126.65
 
AA1317622
View Documents
Alfa Aesar™
1317622
100g
Each for $403.41
 
Description

Description

Specifications

Chemical Identifiers

14175-03-2
750.972
IKPDQYODFRQYAD-UHFFFAOYSA-N
121488066
C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.O.O.O.O.O.O.O.O.O.O.[Sm].[Sm]
C6H26O22Sm2
MFCD00150489
ethanedioic acid,samarium 3+ salt, hydrate 3:2:10, samarium iii oxalate hydrate 2:3:10
oxalic acid;samarium;decahydrate

Specifications

14175-03-2
Sm2(C2O4)3·xH2O
UN3288
IKPDQYODFRQYAD-UHFFFAOYSA-N
oxalic acid;samarium;decahydrate
121488066
Powder
Reacton™
Samarium(III) oxalate hydrate
C6H26O22Sm2
MFCD00150489
ethanedioic acid,samarium 3+ salt, hydrate 3:2:10, samarium iii oxalate hydrate 2:3:10
C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.O.O.O.O.O.O.O.O.O.O.[Sm].[Sm]
750.972
564.78 (Anhydrous)
99.97%
25g
(REO)
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c

H302+H312

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

EINECSNumber : 221-844-8

TSCA : No

Recommended Storage : Ambient temperatures

SDS