missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Samarium(III) oxalate hydrate, REacton™, 99.97% (REO)

Manufacturer:  Alfa Aesar™ 1317622

 View more versions of this product

Catalog No. AA1317622



Specifications

Specifications

Samarium(III) oxalate hydrate
(REO)
Sm2(C2O4)3·xH2O
UN3288
IKPDQYODFRQYAD-UHFFFAOYSA-N
oxalic acid;samarium;decahydrate
121488066
Powder
Reacton™
14175-03-2
C6H26O22Sm2
MFCD00150489
ethanedioic acid,samarium 3+ salt, hydrate 3:2:10
C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.O.O.O.O.O.O.O.O.O.O.[Sm].[Sm]
750.972
564.78 (Anhydrous)
99.97%
100g

Chemical Identifiers

14175-03-2
750.972
IKPDQYODFRQYAD-UHFFFAOYSA-N
121488066
C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.O.O.O.O.O.O.O.O.O.O.[Sm].[Sm]
C6H26O22Sm2
MFCD00150489
ethanedioic acid,samarium 3+ salt, hydrate 3:2:10
oxalic acid;samarium;decahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c

H302+H312

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

EINECSNumber : 221-844-8

TSCA : No

Recommended Storage : Ambient temperatures

Documents