Please login to your online account to display your discounted pricing

Alfa Aesar™ Samarium(III) oxalate decahydrate, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Samarium(III) oxalate decahydrate
CAS 14175-03-2
Linear Formula Sm2(C2O4)3·10H2O
Formula Weight 744.93 (564.78 Anhydrous)
MDL Number MFCD00150489
UN Number UN3288
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1460214

  SDS
alfa aesar™
1460214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1460222

  SDS
alfa aesar™
1460222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 744.93 (564.78 Anhydrous)
Health Hazard 3 GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 14175-03-2
TSCA TSCA
Color White
Health Hazard 1 Warning
Assay Percent Notes (REO)
MDL Number MFCD00150489
Linear Formula Sm2(C2O4)3·10H2O
Quantity 25g
Health Hazard 2 GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
UN Number UN3288
Chemical Name or Material Samarium(III) oxalate decahydrate
Assay 99.9%
Physical Form Crystalline
Safety and Handling

Warning

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.