missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Samarium(III) oxalate decahydrate, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1460222

 View more versions of this product

Catalog No. AA1460222



Specifications

Specifications

Samarium(III) oxalate decahydrate
(REO)
Sm2(C2O4)3·10H2O
UN3288
IKPDQYODFRQYAD-UHFFFAOYSA-N
oxalic acid;samarium;decahydrate
121488066
Crystalline
White
14175-03-2
C6H26O22Sm2
MFCD00150489
samarium iii oxalate hydrate 2:3:10, ethanedioic acid,samarium 3+ salt, hydrate 3:2:10
C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.O.O.O.O.O.O.O.O.O.O.[Sm].[Sm]
750.972
744.93 (564.78 Anhydrous)
99.9%
100g

Chemical Identifiers

14175-03-2
750.972
IKPDQYODFRQYAD-UHFFFAOYSA-N
121488066
C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.C(=O)(C(=O)O)O.O.O.O.O.O.O.O.O.O.O.[Sm].[Sm]
C6H26O22Sm2
MFCD00150489
samarium iii oxalate hydrate 2:3:10, ethanedioic acid,samarium 3+ salt, hydrate 3:2:10
oxalic acid;samarium;decahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

TSCA : TSCA

SDS Documents