missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Samarium(III) nitrate hydrate, REacton™, 99.99% (REO)

$44.68 - $356.80

Chemical Identifiers

CAS 13759-83-6
Molecular Formula H12N3O15Sm
Molecular Weight (g/mol) 444.462
MDL Number MFCD00149857
InChI Key HDCOFJGRHQAIPE-UHFFFAOYSA-N
Synonym 3no3.sm.6h2o, nitric acid, samarium 3+ salt, hexahydrate, samarium 3+ trinitrate hexahydrate, samarium iii nitrate hexahydrate, samarium iii nitrate, hexahydrate 1:3:6, samarium nitrat, samarium nitrat german, samarium nitrate hexahydrate, samarium trinitrate hexahydrate, samariumnitratehexahydrate
PubChem CID 203081
IUPAC Name samarium(3+);trinitrate;hexahydrate
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Sm+3]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1122406
View Documents
Alfa Aesar™
1122406
5g
Each for $44.68
 
AA1122414
View Documents
Alfa Aesar™
1122414
25g
Each for $137.74
 
AA1122422
View Documents
Alfa Aesar™
1122422
100g
Each for $356.80
 
Description

Description

Specifications

Chemical Identifiers

13759-83-6
444.462
HDCOFJGRHQAIPE-UHFFFAOYSA-N
203081
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Sm+3]
H12N3O15Sm
MFCD00149857
3no3.sm.6h2o, nitric acid, samarium 3+ salt, hexahydrate, samarium 3+ trinitrate hexahydrate, samarium iii nitrate hexahydrate, samarium iii nitrate, hexahydrate 1:3:6, samarium nitrat, samarium nitrat german, samarium nitrate hexahydrate, samarium trinitrate hexahydrate, samariumnitratehexahydrate
samarium(3+);trinitrate;hexahydrate

Specifications

13759-83-6
MFCD00149857
3no3.sm.6h2o, nitric acid, samarium 3+ salt, hexahydrate, samarium 3+ trinitrate hexahydrate, samarium iii nitrate hexahydrate, samarium iii nitrate, hexahydrate 1:3:6, samarium nitrat, samarium nitrat german, samarium nitrate hexahydrate, samarium trinitrate hexahydrate, samariumnitratehexahydrate
HDCOFJGRHQAIPE-UHFFFAOYSA-N
samarium(3+);trinitrate;hexahydrate
203081
Crystalline Aggregates
Reacton™
Odorless
Yellow
~3
Samarium(III) nitrate hydrate
H12N3O15Sm
UN1477
Soluble in water and acetonitrile.
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Sm+3]
444.462
336.36 (Anhydrous)
99.99%
2.375 g/mL
Hygroscopic
78°C to 79°C
5g
(REO)
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P380+P375-P372-P380-P501c

H272-H315-H320-H335

DOTInformation : Hazard Class: 5.1; Packaging Group: II

EINECSNumber : 233-798-6

RTECSNumber : VP3150000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS