missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Samarium(III) nitrate hydrate, REacton™, 99.99% (REO)

Manufacturer:  Alfa Aesar™ 1122414

 View more versions of this product

Catalog No. AA1122414



Specifications

Specifications

Samarium(III) nitrate hydrate
(REO)
MFCD00149857
3no3.sm.6h2o
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Sm+3]
444.462
336.36 (Anhydrous)
99.99%
2.375g/mL
Hygroscopic
78°C to 79°C
25g
13759-83-6
H12N3O15Sm
UN1477
HDCOFJGRHQAIPE-UHFFFAOYSA-N
samarium(3+);trinitrate;hexahydrate
203081
Crystalline Aggregates
Reacton™
Odorless
Yellow
~3

Chemical Identifiers

13759-83-6
444.462
HDCOFJGRHQAIPE-UHFFFAOYSA-N
203081
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Sm+3]
H12N3O15Sm
MFCD00149857
3no3.sm.6h2o
samarium(3+);trinitrate;hexahydrate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.

RTECSNumber : VP3150000

TSCA : TSCA

Documents