missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Samarium(III) nitrate hexahydrate, 99.9%, (trace metal basis), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 316150250

 View more versions of this product

Catalog No. AC316150250



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
Specifications

Specifications

Samarium(III) nitrate hexahydrate
13759-83-6
25 g
H12N3O15Sm
MFCD00149857
Solubility in water: soluble
O.O.O.O.O.O.[Sm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
444.46
444.5
Trace Metal Basis
Yellow
Crystalline Powder and/or Crystals
99.9%
Sm(NO3)3·6H2O
samarium nitrate hexahydrate, samarium iii nitrate hexahydrate, samarium trinitrate hexahydrate, samarium nitrat german, samarium iii nitrate, hexahydrate 1:3:6, nitric acid, samarium 3+ salt, hexahydrate, samarium nitrat, samariumnitratehexahydrate, 3no3.sm.6h2o, samarium 3+ trinitrate hexahydrate
HDCOFJGRHQAIPE-UHFFFAOYSA-N
samarium(3+);trinitrate;hexahydrate
203081
99.9%
Plastic bottle

Chemical Identifiers

13759-83-6
444.46
HDCOFJGRHQAIPE-UHFFFAOYSA-N
203081
H12N3O15Sm
MFCD00149857
samarium nitrate hexahydrate, samarium iii nitrate hexahydrate, samarium trinitrate hexahydrate, samarium nitrat german, samarium iii nitrate, hexahydrate 1:3:6, nitric acid, samarium 3+ salt, hexahydrate, samarium nitrat, samariumnitratehexahydrate, 3no3.sm.6h2o, samarium 3+ trinitrate hexahydrate
O.O.O.O.O.O.[Sm+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Safety and Handling

Safety and Handling

GHS H Statement:
May intensify fire; oxidizer.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only