missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Samarium(III) nitrate hexahydrate, 99.9% (REO), Alfa Aesar™

Manufacturer:  Alfa Aesar 1290622*

 View more versions of this product

Catalog No. AA1290622



Specifications

Specifications

Samarium(III) nitrate hexahydrate
99.9% (REO)
MFCD00149857
samarium nitrate hexahydrate, samarium iii nitrate hexahydrate, samarium trinitrate hexahydrate, samarium nitrat german, samarium iii nitrate, hexahydrate 1:3:6, nitric acid, samarium 3+ salt, hexahydrate, samarium nitrat, samariumnitratehexahydrate, 3no3.sm.6h2o, samarium 3+ trinitrate hexahydrate
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Sm+3]
444.462
444.45 (336.36 Anhydrous)
2.375g/cm3 at 20°C
100g
13759-83-6
H12N3O15Sm
Yellow crystals
HDCOFJGRHQAIPE-UHFFFAOYSA-N
samarium(3+);trinitrate;hexahydrate
203081
Crystalline
Odorless

Chemical Identifiers

13759-83-6
444.462
HDCOFJGRHQAIPE-UHFFFAOYSA-N
203081
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Sm+3]
H12N3O15Sm
MFCD00149857
samarium nitrate hexahydrate, samarium iii nitrate hexahydrate, samarium trinitrate hexahydrate, samarium nitrat german, samarium iii nitrate, hexahydrate 1:3:6, nitric acid, samarium 3+ salt, hexahydrate, samarium nitrat, samariumnitratehexahydrate, 3no3.sm.6h2o, samarium 3+ trinitrate hexahydrate
samarium(3+);trinitrate;hexahydrate
Safety and Handling

Safety and Handling

H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

DOTInformation : DOT Class: 5.1, Packaging Group: III

EINECSNumber : 233-798-6

Documents