Please login to your online account to display your discounted pricing

Alfa Aesar™ Samarium(III) isopropoxide, 99% (REO), 5% w/v in toluene/isopropanol

Precursor for CVD and Sol-Gel

Chemical Identifiers

Chemical Name or Material Samarium(III) isopropoxide
CAS 3504-40-3
Linear Formula Sm[OCH(CH3)2]3
Formula Weight 327.5
MDL Number MFCD00058820
UN Number UN3274
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41311AE

  SDS
alfa aesar™
41311AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 327.5
Health Hazard 2 GHS H Statement
H225-H314-H318-H361-H373-H336
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.
Chemical Name or Material Samarium(III) isopropoxide
Physical Form Liquid
Assay 99%
Assay Percent Notes (REO)
Linear Formula Sm[OCH(CH3)2]3
MDL Number MFCD00058820
CAS 3504-40-3
UN Number UN3274
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Concentration 5% w/v in toluene/isopropanol
Quantity 100mL
Odor Alcohol-like
Sensitivity Moisture Sensitive
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLATES SOLUTION, N.O.S.
Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H225-H314-H318-H361-H373-H336
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.