missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Samarium(III) fluoride, anhydrous, REacton™, 99.99% (REO)

Manufacturer:  Alfa Aesar™ 1122604

 View more versions of this product

Catalog No. AA1122604



Specifications

Specifications

Samarium(III) fluoride
13765-24-7
MFCD00049540
Insoluble in water. Moderately soluble in strong mineral acids.
Powder
Reacton™
Hygroscopic
2g
Anhydrous
(REO)
UN3288
207.35
99.99%
Odorless
1306°C

Chemical Identifiers

13765-24-7
MFCD00049540
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

EINECSNumber : 237-367-3

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents