missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Samarium(III) carbonate hydrate, REacton™, 99.99% (REO)

Manufacturer:  Alfa Aesar™ 1317714

 View more versions of this product

Catalog No. AA1317714



Specifications

Specifications

Samarium(III) carbonate hydrate
(REO)
480.73 (Anhydrous)
99.99%
Hygroscopic
25g
38245-37-3
MFCD00150444
Powder
Reacton™
>500°C (decomposition)

Chemical Identifiers

38245-37-3
MFCD00150444
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents