missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Samarium(III) bromide hexahydrate, REacton™, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1460314

 View more versions of this product

Catalog No. AA1460314



Specifications

Specifications

Samarium(III) bromide hexahydrate
Reacton™
(REO)
MFCD00192430
O.O.O.O.O.O.[Br-].[Br-].[Br-].[Sm+3]
498.162
498.15 (390.06 Anhydrous)
Hygroscopic
25g
13517-12-9
99.9%
Br3H12O6Sm
LXJDCZVUWJNXRL-UHFFFAOYSA-K
samarium(3+);tribromide;hexahydrate
57348178
Crystalline
>640°C

Chemical Identifiers

13517-12-9
498.162
LXJDCZVUWJNXRL-UHFFFAOYSA-K
samarium(3+);tribromide;hexahydrate
Br3H12O6Sm
MFCD00192430
57348178
O.O.O.O.O.O.[Br-].[Br-].[Br-].[Sm+3]
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents