Please login to your online account to display your discounted pricing

Alfa Aesar™ Samarium(III) bromide hexahydrate, 99.99%

$248.62 - $248.62

Chemical Identifiers

Chemical Name or Material Samarium(III) bromide hexahydrate
CAS 13517-12-9
Molecular Formula SmBr3·6H2O
Formula Weight 498.15 (390.06 Anhydrous)
MDL Number MFCD00192430
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1819118

  SDS
alfa aesar™
A1819118
50g Each for $248.62
Description & Specifications

Specifications

Formula Weight 498.15 (390.06 Anhydrous)
CAS 13517-12-9
Melting Point >640°C
Quantity 50g
Assay 99.99%
MDL Number MFCD00192430
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Samarium(III) bromide hexahydrate
Health Hazard 1 Warning
Molecular Formula SmBr3·6H2O
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.