Please login to your online account to display your discounted pricing

Alfa Aesar™ Samarium(III) acetate hydrate, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Samarium(III) acetate hydrate
CAS 17829-86-6
Formula Weight 327.48 (Anhydrous)
MDL Number MFCD00150124
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1460414

  SDS
alfa aesar™
1460414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1460422

  SDS
alfa aesar™
1460422
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 327.48 (Anhydrous)
Physical Form Crystalline Aggregates
Assay Percent Notes (REO)
Solubility Information Soluble in water
Density 1.94g/mL
Assay 99.9%
Odor Odorless
Boiling Point 300°C
TSCA TSCA
MDL Number MFCD00150124
CAS 17829-86-6
Sensitivity Hygroscopic
Chemical Name or Material Samarium(III) acetate hydrate
Quantity 25g