missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Samarium(III) acetate hydrate, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 1460422

 View more versions of this product

Catalog No. AA1460422



Specifications

Specifications

Samarium(III) acetate hydrate
(REO)
Soluble in water
Crystalline Aggregates
1.94g/mL
Hygroscopic
100g
17829-86-6
MFCD00150124
327.48 (Anhydrous)
99.9%
Odorless
300°C

Chemical Identifiers

17829-86-6
MFCD00150124
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents