missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Samarium foil, 0.1mm (0.004 in.) thick, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 12421FF

 View more versions of this product

Catalog No. AA12421FF



Description

Description

  • Packaged under argon
Specifications

Specifications

Samarium foil
25 x 25mm
7440-19-9
Mylar under Argon
Sm
UN3178
KZUNJOHGWZRPMI-UHFFFAOYSA-N
samarium
23951
Foil
≈0.47g/25x25mm
0.1mm (0.004 in.) thick
Odorless
(REO)
MFCD00011233
samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
[Sm]
150.36
CHEBI:33374
99.9%

Chemical Identifiers

7440-19-9
150.36
KZUNJOHGWZRPMI-UHFFFAOYSA-N
23951
samarium
Sm
MFCD00011233
samario, unii-42od65l39f, samarium, elemental, atom, powder, ingot, foil, 3n, chips, 3n, ingot, 3n, acmc-1bk0l
CHEBI:33374
[Sm]
Safety and Handling

Safety and Handling

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

TSCA : TSCA

storageNote1 :

  • Packaged under argon

Documents