Please login to your online account to display your discounted pricing

Samarium fluoride, 99.9%, (trace metal basis), ACROS Organics™


Quantity 25g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  299760250

 View more versions of this product

Catalog No. AC299760250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Fine Crystalline Powder
Formula Weight 207.35
Purity Grade Trace Metal Basis
Quantity 25g
Color Slightly yellow
TSCA TSCA
EINECS Number 237-367-3
Assay Percent Range 99.9%
Name Note 99.9%
MDL Number MFCD00049540
Total Trace Metal Impurities 0.1% max.
Packaging Plastic bottle
Chemical Name or Material Samarium fluoride
Linear Formula SmF3
CAS 13765-24-7
Molecular Formula F3Sm
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Samarium fluoride
Name Note 99.9%
CAS 13765-24-7
Purity Grade Trace Metal Basis
Assay Percent Range 99.9%
Molecular Formula F3Sm
Linear Formula SmF3
Formula Weight 207.35
MDL Number MFCD00049540
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents