Please login to your online account to display your discounted pricing

Alfa Aesar™ Samarium Cobalt powder, -30 mesh

Magnet Alloy Additive

Chemical Identifiers

Chemical Name or Material Samarium Cobalt powder
MDL Number MFCD02091682
UN Number UN3089
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1260818

  SDS
alfa aesar™
1260818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1260830

  SDS
alfa aesar™
1260830
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 50g
Chemical Name or Material Samarium Cobalt powder
Health Hazard 2 GHS H Statement
H334-H228-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable solid.
May cause an allergic skin reaction.
Mesh Size −30 Mesh
UN Number UN3089
Avg. Mol. Wt. or Mol. Wt. Range Approx. ratio: Sm:Cu:Fe:Zr:Co; 25.5:5:14:3:52.5 wt%
Physical Form Powder
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210-P284a-P261-P280-P342+P311-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
Sensitivity Air & Moisture Sensitive
MDL Number MFCD02091682
Safety and Handling

Danger

GHS P Statement
P210-P284a-P261-P280-P342+P311-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H334-H228-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable solid.
May cause an allergic skin reaction.