Please login to your online account to display your discounted pricing

Samarium AA Standard, 1mL = 1mg Sm (1,000ppm Sm)Sm2O3 in 3% HNO3, Ricca Chemical


Quantity 100mL
Packaging Poly natural

Manufacturer: ricca chemical company  RIASM1KN100

 View more versions of this product

Catalog No. ASM1KN100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Color Clear colorless
Odor Odorless
Quantity 100mL
Chemical Name or Material Samarium AA Standard
CAS 7732-18-5
CAS Max % 4.68
CAS Min % 0.11
CAS Max % 97.0
Shelf Life 24 months
CAS Min % 4.5
CAS Max % 0.12
CAS 7697-37-2
CAS Min % 93.0
CAS 12060-58-1
Concentration or Composition (by Analyte or Components) 1mL = 1mg Sm (1, 000ppm Sm)Sm2O3 in 3% HNO3
Packaging Poly natural

Specifications

Chemical Name or Material Samarium AA Standard
CAS 7732-18-5
CAS Min % 93.0
CAS Max % 97.0
CAS 7697-37-2
CAS Min % 4.5
CAS Max % 4.68
CAS 12060-58-1
CAS Min % 0.11
CAS Max % 0.12
Accessories

24 months