Please login to your online account to display your discounted pricing

Salsolinol-1-carboxylic acid, Tocris

Endogenous amino acid

$126.32 - $534.65

Chemical Identifiers

Chemical Name or Material Salsolinol-1-carboxylic acid
Assay Percent Range >98%
Molecular Formula C11H13NO4
Formula Weight 223.23
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

063650

 
r & d systems
RDS063650
50mg Each for $534.65

063610

 
r & d systems
RDS063610
10mg Each for $126.32
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 223.23
Assay Percent Range >98%
CAS 57256-34-5
Molecular Formula C11H13NO4
Chemical Name or Material Salsolinol-1-carboxylic acid
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature