Please login to your online account to display your discounted pricing

Salmeterol xinafoate, Tocris™

Long-acting β2 agonist; bronchodilator

$168.15 - $697.40

Chemical Identifiers

Chemical Name or Material Salmeterol xinafoate
Assay Percent Range >99%
Molecular Formula C25H37NO4·C11H8O3
Formula Weight 603.75
Synonym 4-Hydroxy-α1-[[[6-(4-phenylbutoxy)hexyl]amino]methyl]-1, 3-benzenedimethanol 1-hydroxy-2-naphthalenecarboxylic acid salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

471250

 
r & d systems
RDS471250
50mg Each for $697.40

471210

 
r & d systems
RDS471210
10mg Each for $168.15
Description & Specifications

Specifications

Formula Weight 603.75
Quantity 50mg
Chemical Name or Material Salmeterol xinafoate
Assay Percent Range >99%
Molecular Formula C25H37NO4·C11H8O3
CAS 94749-08-3
Synonym 4-Hydroxy-α1-[[[6-(4-phenylbutoxy)hexyl]amino]methyl]-1, 3-benzenedimethanol 1-hydroxy-2-naphthalenecarboxylic acid salt
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature