missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tocris™ Salirasib

Ras inhibitor; also induces autophagy

$97.76 - $410.62

Specifications

Product Type Salirasib
Inhibitors Salirasib
Molecular Weight 358.54
Purity >98
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
498910R Tocris™
RDS498910
10mg Each for $97.76
498950R Tocris™
RDS498950
50mg Each for $410.62
Specifications

Description

Inhibitor of active Ras proteins. Displaces active Ras from the plasma membrane; impairs downstream signaling and inhibits proliferation of endometrial carcinoma cells. Also facilitates Ras degradation. Shown to induce autophagy in several human cancer cell lines.
  • Chemical Name: 2-[[(2E,6E)-3,7,11-Trimethyl-2,6,10-dodecatrien-1-yl]thio]benzoic acid
  • Formula: C22H30O2S
  • CAS Number: 162520-00-5
  • Solubility: Soluble to 100mM in DMSO and to 50mM in 1eq. NaOH
  • Purity: >98
Description & Specifications

Specifications

Salirasib
Salirasib
358.54
>98