Please login to your online account to display your discounted pricing

Alfa Aesar™ Salinomycin, 90%

Chemical Identifiers

Chemical Name or Material Salinomycin
CAS 53003-10-4
Formula Weight 751.00g/mol
MDL Number MFCD25541652
UN Number UN3462
Merck Index 14,8336
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66057LB0

 
alfa aesar™
J66057-LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 5mg
Color White
Melting Point 113°C
Quantity 5mg
Chemical Name or Material Salinomycin
CAS 53003-10-4
Physical Form Powder
Assay 90%
Formula Weight 751.00g/mol
MDL Number MFCD25541652
UN Number UN3462
Merck Index 14,8336
Solubility Information Soluble in methanol; Insoluble in water
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300
Fatal if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packaging Group: II; Proper Shipping Name: TOXINS, EXTRACTED FROM LIVING SOURCES, SOLID, N.O.S.
RTECS Number VO8620000

Applications
Salinomycin was used as a standard in the development of LC-MS/MS method for detection of residual coccidiostats in egg and muscle samples. It was used to screen out CD44+CD24- mesenchymal-like subpopulation within breast carcinomas.

Solubility
Soluble in methanol. Insoluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300
Fatal if swallowed.

2° to 8°C