missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Saline-Sodium Phosphate-EDTA (SSPE), 20X Solution, Fisher BioReagents

Manufacturer:  Fisher Scientific Company BP13284

 View more versions of this product

Catalog No. BP13284


 

Specifications Description & Specifications

Specifications

Saline-Sodium Phosphate-EDTA
7647-14-5,7558-80-7,60-00-4,7732-18-5
2.9 to 3.1M
SSPE Buffer
Poly CUBE
Filtered through a 2-5 micron filter.
4L
20X Solution
Liquid
DNase free
Sodium Chloride (17.5%), Sodium Biphosphate (2.8%), and Ethylenediamine Tetraacetic Acid (0.6%).
Protease free
Clear

Chemical Identifiers

7647-14-5,7558-80-7,60-00-4,7732-18-5
SSPE Buffer
Saline-Sodium Phosphate-EDTA, 20X Solution

Safety and Handling

SDS & Safety Documents

Recommended Storage : RT