Please login to your online account to display your discounted pricing

Saline-Sodium Phosphate-EDTA (SSPE), 20X Solution, Fisher BioReagents


Quantity 20L
Packaging Poly CUBE

Manufacturer: fisher scientific company  BP132820

 View more versions of this product

Catalog No. BP1328-20

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 20L
Filtered Through Filtered through a 2-5 micron filter.
Color Clear
Physical Form Liquid
Concentration 2.9 to 3.1M
DNase DNase free
Protease Protease free
CAS 7732-18-5
Name Note 20X Solution
CAS 7647-14-5
ChemAlert Storage Symbol Gray
Concentration or Composition (by Analyte or Components) Sodium Chloride (17.5%), Sodium Biphosphate (2.8%), and Ethylenediamine Tetraacetic Acid (0.6%).
Packaging Poly CUBE
Recommended Storage RT
Chemical Name or Material Saline-Sodium Phosphate-EDTA
Synonym SSPE Buffer
CAS 7558-80-7
CAS 60-00-4

Specifications

Chemical Name or Material Saline-Sodium Phosphate-EDTA
Name Note 20X Solution
CAS 7647-14-5
CAS 7558-80-7
CAS 60-00-4
CAS 7732-18-5
Synonym SSPE Buffer
Accessories
 

RT

SDS & Safety Documents