Please login to your online account to display your discounted pricing

Saline-Sodium Citrate (SSC), 20X Solution (Molecular Biology), Fisher BioReagents


Quantity 4L
Packaging Poly CUBE

Manufacturer: fisher scientific company  BP13254

 View more versions of this product

Catalog No. BP13254

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Molecular Biology Grade
Quantity 4L
Physical Form Liquid
Filtered Through Filtered through a 2-5 micron filter.
pH 7
Color Clear
Conductivity [NULL]
DNase DNase free
Protease Protease free
Chemical Name or Material Saline-Sodium Citrate
CAS 7732-18-5
Concentration 2.9 to 3.1M
CAS 6132-04-3
Identification Pass Test
Packaging Poly CUBE
CAS 7647-14-5
Name Note 20X Solution
ChemAlert Storage Symbol Gray
Synonym SSC Buffer
Concentration or Composition (by Analyte or Components) Sodium Chloride (17.5%) and Sodium Citrate Dihydrate (8.8%).
CAS 68-04-2
Recommended Storage RT

Specifications

Chemical Name or Material Saline-Sodium Citrate
Name Note 20X Solution
CAS 6132-04-3
CAS 68-04-2
CAS 7647-14-5
CAS 7732-18-5
Purity Grade Molecular Biology Grade
Synonym SSC Buffer
Accessories
 

RT

SDS & Safety Documents