Please login to your online account to display your discounted pricing

Alfa Aesar™ Salicylsalicylic acid, 98%

$213.96 - $824.20

Chemical Identifiers

Chemical Name or Material Salicylsalicylic acid
CAS 552-94-3
Molecular Formula C14H10O5
Formula Weight 258.23
MDL Number MFCD00020252
Synonym 2-Hydroxybenzoic acid 2-carboxyphenyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2244422

  SDS
alfa aesar™
B2244422
100g Each for $213.96

AAB2244436

  SDS
alfa aesar™
B2244436
500g Each for $824.20
Description & Specifications

Specifications

Formula Weight 258.23
Melting Point 132° to 138°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Synonym 2-Hydroxybenzoic acid 2-carboxyphenyl ester
Quantity 100g
Assay 98%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00020252
CAS 552-94-3
Molecular Formula C14H10O5
Health Hazard 1 Warning
RTECS Number DH2080000
Chemical Name or Material Salicylsalicylic acid

Applications
Salicylsalicylic acid is used in organic synthesis and functions as a plant hormone, as a key ingredient in topical anti-acne products.

Solubility
Hydrolyzes slowly in water.

Notes
Moisture Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.