Please login to your online account to display your discounted pricing

Alfa Aesar™ Salicylidene salicylhydrazide, 97%

$62.68 - $204.92

Chemical Identifiers

Chemical Name or Material Salicylidene salicylhydrazide
CAS 3232-36-8
Molecular Formula C14H12N2O3
Formula Weight 256.26
MDL Number MFCD00043494
Beilstein 2944701
Synonym 2-Hydroxybenzylidene 2-hydroxybenzhydrazide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0823506

  SDS
alfa aesar™
L0823506
5g Each for $62.68

AAL0823514

  SDS
alfa aesar™
L0823514
25g Each for $204.92
Description & Specifications

Specifications

Formula Weight 256.26
Chemical Name or Material Salicylidene salicylhydrazide
Molecular Formula C14H12N2O3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
MDL Number MFCD00043494
CAS 3232-36-8
Beilstein 2944701
Synonym 2-Hydroxybenzylidene 2-hydroxybenzhydrazide
Health Hazard 1 Warning
Melting Point 280° to 284°C
Quantity 5g
Assay 97%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.