Please login to your online account to display your discounted pricing

Salicylic Acid (Crystalline/USP), Fisher Chemical

$293.33 - $561.53

Chemical Identifiers

Chemical Name or Material Salicylic Acid
CAS 69-72-7
Purity Grade USP
Physical Form Solid
Assay 99.5 to 101.0% (USP)
Molecular Formula C7H6O3
Linear Formula 2-HOC6H4COOH
Formula Weight 138.12
MDL Number MFCD00002439
Synonym 2-Hydroxybenzoic Acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

A275-212

  SDS
fisher scientific company
FLA275212
Poly Pail 2.5kg Each for $561.53

A275-500

  SDS
fisher scientific company
FLA275500
Poly Bottle 500g Each for $293.33
Case of 6 Each for $1,759.98
Description & Specifications

Specifications

Formula Weight 138.12
Melting Point 158°C
Boiling Point 211°C
Quantity 2.5kg
Assay 99.5 to 101.0% (USP)
Physical Form Solid
Purity Grade USP
pH 2.4
Color Off-white
Vapor Pressure 0.3mbar at 95°C
Residual Solvents Meets Requirements (USP)
Ignition Residue 0.05% max. (USP)
Total Impurity 0.2% max. (USP)
Phenol Relative Retention Time 0.02% max. (USP)
Heavy Metals (as Pb) 20ppm max. (USP)
Packaging Poly Pail
Melting Temp_Begin 158°C min. (USP)
Salicylic Acid Related Compound A 0.1% max. (USP)
Health Hazard 1 SIGNAL WORD: DANGER!
ChemAlert Storage Symbol Gray
Sulfate 0.02% max. (USP)
MDL Number MFCD00002439
Synonym 2-Hydroxybenzoic Acid
Loss on Drying 0.5% max. (USP)
Identification Pass Test (USP)
Other Impurities 0.05% max. (USP)
Chloride 0.014% max. (USP)
Melting Temp_End 161°C max. (USP)
Health Hazard 3 Emergency Overview
Harmful if swallowed. Irritating to eyes, respiratory system and skin.
NFPA
Health: 2
Flammability: 1
Instability: 0
Salicylic Acid Related Compound B 0.05% max.
Chemical Name or Material Salicylic Acid
Molecular Formula C7H6O3
CAS 69-72-7
Linear Formula 2-HOC6H4COOH
Safety and Handling
 

SIGNAL WORD: DANGER!

Emergency Overview
Harmful if swallowed. Irritating to eyes, respiratory system and skin.
NFPA
Health: 2
Flammability: 1
Instability: 0