missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Salicylic acid, ACS, 99+%

Manufacturer:  Alfa Aesar™ 307820E

 View more versions of this product

Catalog No. AA307820E



Specifications

Specifications

Salicylic acid
C7H6O3
774890
salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone
Soluble in alcohol, acetone, ether; Slightly soluble in cold water; More soluble in boiling water; Water solubility increased by Na3PO4, alkali acetates or alkali citrates
C1=CC=C(C(=C1)C(=O)O)O
138.122
CHEBI:16914
Powder
ACS
211°C (20mmHg)
157° (315°F)
69-72-7
MFCD00002439
14,8332
1.443g/mL
YGSDEFSMJLZEOE-UHFFFAOYSA-N
2-hydroxybenzoic acid
338
138.12
≥99%
158°C to 160°C
2500g

Chemical Identifiers

69-72-7
138.122
YGSDEFSMJLZEOE-UHFFFAOYSA-N
338
2-hydroxybenzoic acid
C7H6O3
MFCD00002439
salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone
CHEBI:16914
C1=CC=C(C(=C1)C(=O)O)O
Safety and Handling

Safety and Handling

GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : VO0525000

TSCA : TSCA

SDS Documents