Learn More
Salicylhydroxamic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 132620050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Salicylhydroxamic acid | |
89-73-6 | |
Beige-Pink | |
98.5% min. (HPLC) | |
C7H7NO3 | |
5 g | |
15,8468 | |
Solubility in water: slightly soluble. Other solubilities: very soluble in ethanol, diethyl ether, soluble with alkalies | |
C1=CC=C(C(=C1)C(=O)NO)O | |
153.14 | |
CHEBI:45615 | |
99% |
99% | |
177.0°C to 185.0°C | |
Authentic | |
Glass bottle | |
HOC6H4C(=NOH)OH | |
10,98 | |
salicylhydroxamic acid, 2-hydroxybenzohydroxamic acid, salicylohydroxamic acid, sham, salicylohydroximic acid, benzamide, n,2-dihydroxy, 2-hydroxybenzhydroxamic acid, o-hydroxybenzohydroxamic acid, sha, 2,n-dihydroxy-benzamide | |
HBROZNQEVUILML-UHFFFAOYSA-N | |
N,2-dihydroxybenzamide | |
66644 | |
153.14 | |
Fine Crystalline Powder |
Chemical Identifiers
89-73-6 | |
153.14 | |
salicylhydroxamic acid, 2-hydroxybenzohydroxamic acid, salicylohydroxamic acid, sham, salicylohydroximic acid, benzamide, n,2-dihydroxy, 2-hydroxybenzhydroxamic acid, o-hydroxybenzohydroxamic acid, sha, 2,n-dihydroxy-benzamide | |
CHEBI:45615 | |
C1=CC=C(C(=C1)C(=O)NO)O |
C7H7NO3 | |
HBROZNQEVUILML-UHFFFAOYSA-N | |
66644 | |
N,2-dihydroxybenzamide |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing genetic defects.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 201-934-3
RTECSNumber : VO6870000
RUO – Research Use Only