Please login to your online account to display your discounted pricing

Salicylhydrazide 98%, ACROS Organics™


Quantity 25g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  132610250

 View more versions of this product

Catalog No. AC132610250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 152.15
Quantity 25g
Color White to off-white
Beilstein 10, 100
TSCA TSCA
RTECS Number VO3700000
EINECS Number 213-311-3
Melting Point 147° to 150°C
Infrared Spectrum Authentic
Chemical Name or Material Salicylhydrazide
Name Note 98%
Assay Percent Range 98%
Linear Formula 2-(HO)C6H4CONHNH2
MDL Number MFCD00007599
CAS 936-02-7
Synonym Salicylic acid hydrazide
Packaging Plastic bottle
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C7H8N2O2

Specifications

Chemical Name or Material Salicylhydrazide
Name Note 98%
CAS 936-02-7
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H8N2O2
Linear Formula 2-(HO)C6H4CONHNH2
Formula Weight 152.15
MDL Number MFCD00007599
Synonym Salicylic acid hydrazide
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents