Please login to your online account to display your discounted pricing

Alfa Aesar™ Salicylanilide, 98%

$103.82 - $411.35

Chemical Identifiers

Chemical Name or Material Salicylanilide
CAS 87-17-2
Molecular Formula C13H11NO2
Formula Weight 213.24
MDL Number MFCD00002212
Merck Index 14,8330
Synonym 2-Hydroxybenzanilide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA103350E

  SDS
alfa aesar™
A103350E
2500g Each for $411.35

AAA1033536

  SDS
alfa aesar™
A1033536
500g Each for $103.82
Description & Specifications

Specifications

Formula Weight 213.24
Chemical Name or Material Salicylanilide
TSCA TSCA
Assay 98%
Molecular Formula C13H11NO2
MDL Number MFCD00002212
CAS 87-17-2
Merck Index 14,8330
Synonym 2-Hydroxybenzanilide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 136° to 138°C
Quantity 2500g

Applications
It is employed as a drug used to treat rheumatoid arthritis and has principally analgesic, antipyretic and anti-inflammatory actions. Non-narcotic analgesics do not bind to opioid receptors. It is used mainly in medicated soaps.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.