missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Salicylaldoxime, 98%, ACROS Organics™

Manufacturer:  Acros Organics 207400250

 View more versions of this product

Catalog No. AC207400250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(1% soln. in chloroform) clear
Authentic
0.1% max.
Bordeaux to Purple-Red
25g
98%
2-(HO)C6H4CH=NOH
08, 49
2-1e-hydroxyimino methyl phenol
C1=CC(=CNO)C(=O)C=C1
137.14
137.14
≥97.5% (GC)
Salicylaldoxime
Glass bottle
0.5% max. (K.F.)
56°C to 59°C
94-67-7
C7H7NO2
MFCD00002120
15, 8464
GFCNBJDXQMZOOC-UHFFFAOYSA-N
6-[(hydroxyamino)methylidene]cyclohexa-2,4-dien-1-one
6740756
Powder

Chemical Identifiers

94-67-7
137.14
GFCNBJDXQMZOOC-UHFFFAOYSA-N
6740756
C1=CC(=CNO)C(=O)C=C1
C7H7NO2
MFCD00002120
2-1e-hydroxyimino methyl phenol
6-[(hydroxyamino)methylidene]cyclohexa-2,4-dien-1-one
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents