Please login to your online account to display your discounted pricing

Alfa Aesar™ Salicylaldehyde hydrazone, 98%

$42.50 - $412.85

Chemical Identifiers

Chemical Name or Material Salicylaldehyde hydrazone
CAS 3291-00-7
Molecular Formula C7H8N2O
Formula Weight 136.15
MDL Number MFCD00007626
Synonym 2-Hydroxybenzaldehyde hydrazone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2287806

  SDS
alfa aesar™
B2287806
5g Each for $42.50

AAB2287814

  SDS
alfa aesar™
B2287814
25g Each for $119.93

AAB2287822

  SDS
alfa aesar™
B2287822
100g Each for $412.85
Description & Specifications

Specifications

Formula Weight 136.15
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 95° to 98°C
Synonym 2-Hydroxybenzaldehyde hydrazone
MDL Number MFCD00007626
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Molecular Formula C7H8N2O
Assay 98%
Chemical Name or Material Salicylaldehyde hydrazone
CAS 3291-00-7
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.