missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Salicylaldehyde, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1383322

 View more versions of this product

Catalog No. AAA1383322


 

Description

Description

Applications
Precursor to a variety of chelating agents.Salicylaldehyde is used as a precursor to prepare catechol, benzofuran, salicylaldehydimine and 3-carbethoxycoumarin. It is used for colorimetric assay of isoleucine as well as utilized for the determination of primary amine by fluorescence method. It is a key starting material to a various chelating agents. It is used as flavor and fragrance components. Its derivative salicylaldehyde thiosemicarbazone is used for the simultaneous determination of molybdenum and iron by spectrophotometric method.

Solubility
Miscible with alcohol, ether, acetone, benzene and most organic solvents. Slightly miscible with water.

Notes
Incompatible with acids, strong reducing agents, strong oxidizing agents, strong bases and halogens.
Specifications

Specifications

Salicylaldehyde
1.168
99%
MFCD00003317
14,8326
C1=CC=C(C(=C1)C=O)O
122.123
CHEBI:16008
Liquid
90-02-8
99%
C7H6O2
471388
SMQUZDBALVYZAC-UHFFFAOYSA-N
2-hydroxybenzaldehyde
6998
122.12
100g

Chemical Identifiers

90-02-8
122.123
SMQUZDBALVYZAC-UHFFFAOYSA-N
CHEBI:16008
C7H6O2
MFCD00003317
6998
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335-H227

GHS P Statement
P210-P261-P280-P305+P351+P338-P304+P340-P362-P301+P312-P312-P405-P501a

Warning

RTECSNumber : VN5250000

TSCA : TSCA

SDS Documents